Anti-PP1CG Rabbit Polyclonal Antibody (Cy7®)

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-5422R-CY7
BOSSBS-5422R-CY7EA 900 EUR
BOSSBS-5422R-CY7
Anti-PP1CG Rabbit Polyclonal Antibody (Cy7®)
Antilichamen
Protein phosphatase 1 (PP1) is essential for cell division, and participates in the regulation of glycogen metabolism, muscle contractility and protein synthesis. The protein is involved in regulation of ionic conductances and long term synaptic plasticity. It may play an important role in dephosphorylating substrates such as the postsynaptic density associated Ca (2+)/calmodulin dependent protein kinase II.PP1 comprises a catalytic subunit, PPP1CA, PPP1CB or PPP1CC (PP1C gamma), which is folded into its native form by inhibitor 2 and glycogen synthetase kinase 3, and then complexed to oneMore Product Information
Order Now

Specificaties

antigeen symbool PP1CG
Environmentally Preferable
Type antilichaam Primair,Serine/threonine protein phosphatase PP1-gamma catalytic subuni
Antigeen synoniemen Protein phosphatase 1C catalytic subunit|PP-1G|Serine/threonine protein phosphatase PP1-gamma catalytic subunit
Type antilichaam Primair,Serine/threonine protein phosphatase PP1-gamma catalytic subuni
Clonality Polyklonaal
Conjugatie Cy7®
Reactivity Human,Rat,Mouse
Host Rabbit
Gen-ID 5501
Isotype IgG
ImmunoChemie Yes
Formulier liquid
Purification Purified by Protein A
Concentratie 1 μg/μl
Kruisadsorptie No
Opslagbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Opslagtemperatuur Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR