Anti-SLC29A4 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-5637R-A555
BOSSBS-5637R-A555EA 1350 EUR
BOSSBS-5637R-A555
Anti-SLC29A4 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antilichamen
SLC29A4 (Solute carrier family 29 (nucleoside transporters), member 4) is involved in the re-uptake of monoamines into presynaptic neurons, thus determining the intensity and duration of monoamine neural signaling. It transports serotonin, dopamine and the neurotoxin 1-methyl-4-phenylpyridinium. It may also contribute to the regulation of extracellular adenosine concentrations.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SLC29A4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Specificaties

antigeen symbool SLC29A4
Environmentally Preferable
Type antilichaam Primair,Solute Carrier Family 29 Member A4
Antigeen synoniemen ENT4|PSEC0113|Solute carrier family 29 member 4|hENT4|Plasma membrane monoamine transporter|Equilibrative nucleoside transporter 4|PMAT|SLC29A4
Type antilichaam Primair,Solute Carrier Family 29 Member A4
Clonality Polyklonaal
Conjugatie Alexa Fluor® 555
Reactivity Human,Rat,Mouse
Host Rabbit
Gen-ID 222962
Isotype IgG
ImmunoChemie Yes
Modification phospho-Tyr198
Formulier liquid
Purification Purified by Protein A
Concentratie 1 μg/μl
Kruisadsorptie No
Opslagbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Opslagtemperatuur Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR