Anti-AZU1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-5736R-HRP
BOSSBS-5736R-HRPEA 900 EUR
BOSSBS-5736R-HRP
Anti-AZU1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antilichamen
TACC2 is a conserved family of centrosome and microtubule interacting proteins that are implicated in cancer. This gene encodes a protein that concentrates at centrosomes throughout the cell cycle. This gene lies within a chromosomal region associated with tumorigenesis. Expression of this gene is thought to affect the progression of breast tumors. Expression of this gene is also induced by erythropoietin.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: AZU1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR