Anti-ACVR1C Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-5777R-HRP
BOSSBS-5777R-HRPEA 900 EUR
BOSSBS-5777R-HRP
Anti-ACVR1C Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antilichamen
Serine/threonine protein kinase which forms a receptor complex on ligand binding. The receptor complex consisting of 2 type II and 2 type I transmembrane serine/threonine kinases. Type II receptors phosphorylate and activate type I receptors which autophosphorylate, then bind and activate SMAD transcriptional regulators, SMAD2 and SMAD3. Receptor for activin AB, activin B and NODAL. Plays a role in cell differentiation, growth arrest and apoptosis.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: ACVR1C
Clonality: Polyclonal
Clone:
Conjugation: HRP (More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR