Anti-RNF139 Rabbit Polyclonal Antibody (Cy5®)
Leverancier: Bioss
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BS-6096R-CY5
BOSSBS-6096R-CY5EA 900 EUR
BOSSBS-6096R-CY5
Anti-RNF139 Rabbit Polyclonal Antibody (Cy5®)
Antilichamen
The protein encoded by this gene is a multi-membrane spanning protein containing a RING-H2 finger. This protein is located in the endoplasmic reticulum, and has been shown to possess ubiquitin ligase activity. This gene was found to be interrupted by a t(3:8) translocation in a family with hereditary renal and non-medulary thyroid cancer. Studies of the Drosophila counterpart suggested that this protein may interact with tumor suppressor protein VHL, as well as with COPS5/JAB1, a protein responsible for the degradation of tumor suppressor CDKN1B/P27KIP.
Recommended Dilutions: IF(IHC-P): 1:More Product Information