Anti-OTUB Rabbit Polyclonal Antibody (Cy5.5®)

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-6236R-CY5.5
BOSSBS-6236R-CY5.5EA 900 EUR
BOSSBS-6236R-CY5.5
Anti-OTUB Rabbit Polyclonal Antibody (Cy5.5®)
Antilichamen
This gene encodes one of several deubiquitylating enzymes. Ubiquitin modification of proteins is needed for their stability and function; to reverse the process, deubiquityling enzymes remove ubiquitin. This protein contains an OTU domain and binds Ubal (ubiquitin aldehyde); an active cysteine protease site is present in the OTU domain.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: OTUB
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR