Anti-TLX3 Rabbit Polyclonal Antibody

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-6240R
BOSSBS-6240REA 725 EUR
BOSSBS-6240R
Anti-TLX3 Rabbit Polyclonal Antibody
Antilichamen
TLX3 (HOX11L2, RNX) belongs to a family of orphan homeobox genes that encode DNA binding nuclear transcription factors. Members of the HOX11 gene family are characterized by a threonine 47 replacing cytosine in the highly conserved homeodomain. This transcription factor is required for hematopoiesis. It is overexpressed in childhood acute lumphoblastic leukemia.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: TLX3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR