Anti-CH25H Rabbit Polyclonal Antibody (Cy3®)

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-6480R-CY3
BOSSBS-6480R-CY3EA 915 EUR
BOSSBS-6480R-CY3
Anti-CH25H Rabbit Polyclonal Antibody (Cy3®)
Antilichamen
Catalyzes the formation of 25-hydroxycholesterol from cholesterol, leading to repress cholesterol biosynthetic enzymes. May play an important role in regulating lipid metabolism by synthesizing a corepressor that blocks sterol regulatory element binding protein (SREBP) processing. In testis, production of 25-hydroxycholesterol by macrophages may play a role in Leydig cell differentiation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CH25H
Clonality: Polyclonal
Clone: 0
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR