Anti-OSBP2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Leverancier: Bioss
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BS-6560R-FITC
BOSSBS-6560R-FITCEA 900 EUR
BOSSBS-6560R-FITC
Anti-OSBP2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antilichamen
The Oxysterol-binding protein (OSBP) family of proteins consist of OSBP (OSBP1) and OSBP2 (ORP-4), which share a high overall similarity. OSBPs are involved in lipid metabolism and signal transduction, as well as vesicle transport, and can translocate to the periphery of Golgi membranes when they are bound to oxysterols. The OSBP protein transports sterols from lysosomes to the nucleus, where sterols downregulate the genes for HMG synthetase, HMG-CoA reductase and the low density lipoprotein receptor (LDLR). OSBP localizes to the cytosol and is widely expressed, while OSBP2 is mainlyMore Product Information