Anti-IHH Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-6624R-A680
BOSSBS-6624R-A680EA 893 EUR
BOSSBS-6624R-A680
Anti-IHH Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antilichamen
Ihh is an intercellular signalling peptide essential for a variety of patterning events during development. It binds to the patched (PTC) receptor, which functions in association with smoothened (SMO), to activate the transcription of target genes. Ihh induces the expression of parathyroid hormone-related protein (PTHRP) and is implicated in endochondral ossification, possibly regulating the balance between growth and ossification of the developing bones.

Type: Primary
Antigen: IHH
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 161-260/411
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Specificaties

Type antilichaam Primair,IHH
Environmentally Preferable
Antigeen synoniemen Indian Hedgehog|HHG2|Centromere protein B|CENPB_HUMAN|CENP-B|Major centromere autoantigen B
Type antilichaam Primair,IHH
Clonality Polyklonaal
Conjugatie Alexa Fluor® 680
Reactivity Rat
Host Rabbit
Gen-ID 3549
Isotype IgG
ImmunoChemie Yes
ImmunoFluorescentie Yes
Modification Unmodified
Epitoop 161-260/411
Immunogeen KLH conjugated synthetic peptide derived from human IHH
Formulier Liquid
Moleculair gewicht 45 kDa
Purification Purified by Protein A
Concentratie 1 µg/µl
Opslagbuffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Opslagtemperatuur Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Shipping temperature 4 °C
Tested applications ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR