Anti-Septin5 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Leverancier: Bioss
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BS-6905R-A680
BOSSBS-6905R-A680EA 893 EUR
BOSSBS-6905R-A680
Anti-Septin5 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antilichamen
This gene is a member of the septin gene family of nucleotide binding proteins, originally described in yeast as cell division cycle regulatory proteins. Septins are highly conserved in yeast, Drosophila, and mouse and appear to regulate cytoskeletal organisation. Disruption of septin function disturbs cytokinesis and results in large multinucleate or polyploid cells. This gene is mapped to 22q11, the region frequently deleted in DiGeorge and velocardiofacial syndromes. A translocation involving the MLL gene and this gene has also been reported in patients with acute myeloid leukemia.More Product Information