Anti-PRMT5 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-6992R-FITC
BOSSBS-6992R-FITCEA 900 EUR
BOSSBS-6992R-FITC
Anti-PRMT5 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antilichamen
Arginine methyltransferase that can both catalyze the formation of omega-N monomethylarginine (MMA) and symmetrical dimethylarginine (sDMA), with a preference for the formation of MMA. Specifically mediates the symmetrical dimethylation of arginine residues in the small nuclear ribonucleoproteins Sm D1 (SNRPD1) and Sm D3 (SNRPD3); such methylation being required for the assembly and biogenesis of snRNP core particles. Methylates SUPT5H. Mono- and dimethylates arginine residues of myelin basic protein (MBP) in vitro. Plays a role in the assembly of snRNP core particles. May play a role inMore Product Information
Order Now

Specificaties

antigeen symbool PRMT5
Environmentally Preferable
Type antilichaam Primair,Protein Arginine Methyltransferase 5
Antigeen synoniemen SKB1|HRMT1L5|IBP72|SKB1Hs|JBP1
Type antilichaam Primair,Protein Arginine Methyltransferase 5
Clonality Polyklonaal
Conjugatie FITC (Fluoresceïne-isothiocyanaat)
Reactivity Human,Rat,Mouse
Host Rabbit
Gen-ID 10419
Isotype IgG
ImmunoChemie Yes
Formulier liquid
Purification Purified by Protein A
Concentratie 1 μg/μl
Kruisadsorptie No
Opslagbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Opslagtemperatuur Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR