Anti-ELF1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-7042R-A750
BOSSBS-7042R-A750EA 893 EUR
BOSSBS-7042R-A750
Anti-ELF1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antilichamen
E74 ike Factor 1 is a transcription factor that may be required for the T-cell receptor mediated activation of HIV2 gene expression. It binds to two purine-rich motifs in the HIV-2 enhancer and transcriptionally activates the LYN and mouse BLK promoters.

Type: Primary
Antigen: ELF1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 201-300/619
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR