Anti-p53AIP1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-7094R-A750
BOSSBS-7094R-A750EA 893 EUR
BOSSBS-7094R-A750
Anti-p53AIP1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antilichamen
This gene is specifically expressed in the thymus, and encodes a protein that is localised to the mitochondrion. The expression of this gene is inducible by p53, and it is thought to play an important role in mediating p53-dependent apoptosis. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.

Type: Primary
Antigen: p53AIP1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 65-124/124
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR