Anti-POLD2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Leverancier: Bioss
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BS-7416R-A350
BOSSBS-7416R-A350EA 893 EUR
BOSSBS-7416R-A350
Anti-POLD2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antilichamen
DNA replication, recombination and repair, all of which are necessary for genomic stability, require the presence of exonucleases (1). In DNA replication, these enzymes are involved in the processing of Okazaki fragments, whereas in DNA repair, they function to excise damaged DNA fragments and correct recombinational mismatches (2). These exonucleases include the family of DNA polymerases (3). DNA pol α, β, ∂, and e are involved in DNA replication and repair (4). DNA pol ∂ and DNA pol e are multisubunit enzymes, with DNA pol ∂ consisting of two subunits p125, which interacts with theMore Product Information