Anti-KIF2B Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-7751R-HRP
BOSSBS-7751R-HRPEA 900 EUR
BOSSBS-7751R-HRP
Anti-KIF2B Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antilichamen
KIF2B belongs to the kinesin-like protein family. It is a plus end-directed microtubule-dependent motor required for spindle assembly and chromosome movement. It also has microtubule depolymerization activity.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: KIF2B
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR