Anti-ELFN2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-7809R-A350
BOSSBS-7809R-A350EA 893 EUR
BOSSBS-7809R-A350
Anti-ELFN2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antilichamen
LRRC62 contains 1 fibronectin type III domain and 5 LRR (leucine rich) repeats. The exact function of LRRC62 remains unknown.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ELFN2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR