Anti-UBE2V1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Leverancier: Bioss
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BS-7882R-A680
BOSSBS-7882R-A680EA 893 EUR
BOSSBS-7882R-A680
Anti-UBE2V1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antilichamen
Has no ubiquitin ligase activity on its own. The UBE2V1-UBE2N heterodimer catalyzes the synthesis of non-canonical poly-ubiquitin chains that are linked through Lys-63. This type of poly-ubiquitination activates IKK and does not seem to involve protein degradation by the proteasome. Plays a role in the activation of NF-kappa-B mediated by IL1B, TNF, TRAF6 and TRAF2. Mediates transcriptional activation of target genes. Plays a role in the control of progress through the cell cycle and differentiation. Plays a role in the error-free DNA repair pathway and contributes to the survival of cellsMore Product Information