Anti-CCNT2 Rabbit Polyclonal Antibody (Cy5®)

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-7891R-CY5
BOSSBS-7891R-CY5EA 900 EUR
BOSSBS-7891R-CY5
Anti-CCNT2 Rabbit Polyclonal Antibody (Cy5®)
Antilichamen
Regulatory subunit of the cyclin-dependent kinase pair (CDK9/cyclin T) complex, also called positive transcription elongation factor B (P-TEFB), which is proposed to facilitate the transition from abortive to production elongation by phosphorylating the CTD (carboxy-terminal domain) of the large subunit of RNA polymerase II (RNAP II).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CCNT2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR