Anti-PLK5 Rabbit Polyclonal Antibody (Cy5®)

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-7918R-CY5
BOSSBS-7918R-CY5EA 900 EUR
BOSSBS-7918R-CY5
Anti-PLK5 Rabbit Polyclonal Antibody (Cy5®)
Antilichamen
PLK5 (Serine/threonine-protein kinase PLK5) belongs to the protein kinase superfamily and Ser/Thr protein kinase family and CDC5/Polo subfamily. PLK5P contains 1 POLO box domain and 1 Serine/Threonine protein kinase catalytic domain.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PLK5
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Specificaties

antigeen symbool PLK5
Environmentally Preferable
Type antilichaam Primair,Polo-Like Kinase 5
Antigeen synoniemen inactive serine/threonine-protein kinase PLK5|polo-like kinase 5 pseudogene|PLK-5|SgK384ps|PLK5P
Type antilichaam Primair,Polo-Like Kinase 5
Clonality Polyklonaal
Conjugatie Cy5®
Reactivity Human,Rat,Mouse
Host Rabbit
Isotype IgG
ImmunoChemie Yes
Formulier liquid
Purification Purified by Protein A
Concentratie 1 μg/μl
Kruisadsorptie No
Opslagbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Opslagtemperatuur Store at –20 °C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR