Anti-CCDC25 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Leverancier: Bioss
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BS-8125R-A647
BOSSBS-8125R-A647EA 893 EUR
BOSSBS-8125R-A647
Anti-CCDC25 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antilichamen
CCDC25 is a 208 amino acid protein encoded by a gene that maps to human chromosome 8p21.1. Made up of nearly 146 million bases, chromosome 8 encodes about 800 genes. Translocation of portions of chromosome 8 with amplifications of the c-Myc gene are found in some leukemias and lymphomas, and typically associated with a poor prognosis. Portions of chromosome 8 have been linked to schizophrenia and bipolar disorder. Trisomy 8, also known as Warkany syndrome 2, most often results in early miscarriage but is occasionally seen in a mosaic form in surviving patients who suffer to a varying degreeMore Product Information