Anti-GAREM Rabbit Polyclonal Antibody (Cy7®)

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-8218R-CY7
BOSSBS-8218R-CY7EA 900 EUR
BOSSBS-8218R-CY7
Anti-GAREM Rabbit Polyclonal Antibody (Cy7®)
Antilichamen
Encoding over 300 genes, chromosome 18 contains about 76 million bases. Trisomy 18, or Edwards syndrome, is the second most common trisomy after Downs syndrome. Symptoms of Edwards syndrome include low birth weight, a variety of physical development defects, heart deformations and breathing difficulty. Translocation between chromosome 18 and 14 is the most common translocation in cancers and occurs in follicular lymphomas. Niemann-Pick disease, hereditary hemorrhagic telangiectasia and erythropoietic protoporphyria are associated with chromosome 18. The TGF∫ modulators, Smad2, Smad4 andMore Product Information
Order Now

Specificaties

antigeen symbool GAREM
Environmentally Preferable
Type antilichaam Primair,GRB2 associated, regulator of MAPK1
Antigeen synoniemen Gm944|C18orf11|FAM59A
Type antilichaam Primair,GRB2 associated, regulator of MAPK1
Clonality Polyklonaal
Conjugatie Cy7®
Reactivity Human,Rat,Mouse
Host Rabbit
Gen-ID 64762
Isotype IgG
ImmunoChemie Yes
Formulier liquid
Purification Purified by Protein A
Concentratie 1 μg/μl
Kruisadsorptie No
Opslagbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Opslagtemperatuur Store at –20 °C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR