Anti-CECR6 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-9006R-A488
BOSSBS-9006R-A488EA 893 EUR
BOSSBS-9006R-A488
Anti-CECR6 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antilichamen
Adenosine deaminase is an enzyme that is present in most tissues and exists predominantly as a monomer, although in some tissues it is associated with adenosine deaminase-binding protein. Adenosine deaminase degrades extracellular adenosine, which is toxic for lymphocytes. A novel family of growth factors that share sequence similarity to adenosine deaminase has been identified. The cat eye syndrome critical region protein (CECR) family includes CECR1, CECR2, CECR3, CECR4, CECR5, CECR6, CECR7, CECR8 and CECR9. The genes encoding CECR proteins are candidates for Cat Eye Syndrome (CES), aMore Product Information
Order Now

Specificaties

antigeen symbool CECR6
Environmentally Preferable
Type antilichaam Primair,Cat eye syndrome chromosome region, candidate 6
Antigeen synoniemen Cat eye syndrome chromosome region candidate 6 homolog|Cat eye syndrome chromosome region candidate 6 homolog human|RGD1565302|AW048664|CECR6_HUMAN.|Cat eye syndrome chromosome region candidate 6
Type antilichaam Primair,Cat eye syndrome chromosome region, candidate 6
Clonality Polyklonaal
Conjugatie Alexa Fluor® 488
Reactivity Human,Rat,Mouse
Host Rabbit
Gen-ID 27439
Isotype IgG
ImmunoChemie Yes
Formulier liquid
Purification Purified by Protein A
Concentratie 1 μg/μl
Kruisadsorptie No
Opslagbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Opslagtemperatuur Store at –20 °C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR