Anti-HMBS Rabbit Polyclonal Antibody
Leverancier: Bioss
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BS-9068R
BOSSBS-9068REA 725 EUR
BOSSBS-9068R
Anti-HMBS Rabbit Polyclonal Antibody
Antilichamen
PBGD, also designated hydroxymethylbilane synthase, is a cytoplasmic enzyme found in the heme synthesis pathway. PBGD belongs to the HMBS (hydroxymethylbilane synthase) family. Deficiency of PBGD causes errors in pyrrole metabolism, which in turn leads to an inherited autosomal disorder called acute intermittent porphyria (AIP). AIP is characterized by acute attacks of neurological dysfunctions with hypertension, tachycardia, peripheral neurologic disturbances, abdominal pain and excessive amounts of aminolevulinic acid and porphobilinogen in the urine.
WB(1:100-1000), IHC-P(1:100-500),More Product Information