Anti-HMBS Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Leverancier: Bioss
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BS-9068R-FITC
BOSSBS-9068R-FITCEA 900 EUR
BOSSBS-9068R-FITC
Anti-HMBS Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antilichamen
PBGD, also designated hydroxymethylbilane synthase, is a cytoplasmic enzyme found in the heme synthesis pathway. PBGD belongs to the HMBS (hydroxymethylbilane synthase) family. Deficiency of PBGD causes errors in pyrrole metabolism, which in turn leads to an inherited autosomal disorder called acute intermittent porphyria (AIP). AIP is characterized by acute attacks of neurological dysfunctions with hypertension, tachycardia, peripheral neurologic disturbances, abdominal pain and excessive amounts of aminolevulinic acid and porphobilinogen in the urine.
Recommended Dilutions: IF(IHC-P): 1:More Product Information