Anti-RNF128 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Leverancier: Bioss
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BS-9251R-A647
BOSSBS-9251R-A647EA 893 EUR
BOSSBS-9251R-A647
Anti-RNF128 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antilichamen
The protein encoded by this gene is a type I transmembrane protein that localizes to the endocytic pathway. This protein contains a RING zinc-finger motif and has been shown to possess E3 ubiquitin ligase activity. Expression of this gene in retrovirally transduced T cell hybridoma significantly inhibits activation-induced IL2 and IL4 cytokine production. Induced expression of this gene was observed in anergic CD4(+) T cells, which suggested a role in the induction of anergic phenotype. Alternatively spliced transcript variants encoding distinct isoforms have been reported. [provided byMore Product Information