Contact slides, HYCON®, microbial monitoring of flat and curved surfaces in the food & beverage industry

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

1.44018.0020 1.44019.0020 1.44022.0020 1.44021.0020 1.44023.0100 1.44022.0100
BTSM1.44018.0020EA 112 EUR
BTSM1.44018.0020 BTSM1.44019.0020 BTSM1.44022.0020 BTSM1.44021.0020 BTSM1.44023.0100 BTSM1.44022.0100
Contact slides, HYCON®, microbial monitoring of flat and curved surfaces in the food & beverage industry
Media voor microbiologie Geprepareerde media voor microbiologie

HYCON® Contact Slides for microbial monitoring of flat and curved surfaces are designed as standardised agar media in a flexible foil. Their unique design makes them universal tools for easy microbial monitoring of flat and curved surfaces. The choice of formulations includes total count media as well as selective media for yeasts and moulds, and for coliform organisms. Each contact slide is welded shrink-wrapped into a resealable sleeve to enable individual strips to be safely sterilised a number of times by peroxide gassing.


  • Individually sealed, H₂O₂-impermeable primary packaging
  • Resealable sleeve ensures sterility and enables safe handling and transportation
  • Produced in clean rooms, rigorously quality controlled, including 100% visual inspection of each contact slide
  • Growth promotion, packaging and storage extensively validated
  • Standardised contact area meets international guidelines (25 cm²)
  • Can be used for flat, curved or irregular surfaces

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR