Experimentation kits, cleavage of Lambda DNA with Eco RI endonuclease: intro to restriction enzymes
Leverancier: EDVOTEK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
212
EDVO212EA 153 EUR
EDVO212
Experimentation kits, cleavage of Lambda DNA with Eco RI endonuclease: intro to restriction enzymes
Educatief materiaal Educatief materiaal over biologie Leeractiviteiten voor biochemie
The DNA from bacteriophage Lambda is a well-characterised linear molecule containing six recognition sites for Eco RI (5 distinct sites, 2 are very close in size). In this experiment, Lambda DNA is digested by the Eco RI endonuclease. The digestion products are analysed by agarose gel electrophoresis.
  • For 5 lab groups
  • Complete in 90 minutes
Also required: Electrophoresis tank, power supply, 5 to 50 µl and 40 µl fixed volume micropipettes, water bath, and white light box.
Leveringsinformatie: Kit includes instructions, Lambda DNA, Dryzymes™,More Product Information