β-Nicotinamide adenine dinucleotid, oxidised form (NAD, oxidised form) ≥93%

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

0210031905 0210031901 0210031925
ICNA0210031905EA 204 EUR
ICNA0210031905 ICNA0210031901 ICNA0210031925
β-Nicotinamide adenine dinucleotid, oxidised form (NAD, oxidised form) ≥93%
β-Nicotinamide adenine dinucleotid, oxidised form (NAD, oxidized form)

This grade of NAD is intended for applications where maximum reagent purity is not critical. Well suited for use on most automatic analysers.


  • White powder, soluble in water (200 mg/ml)
  • pH: 2 - 4 (1% aqueous solution)


Solutions should be aliquoted and stored at –20 °C for up to one week.

Formule: C₂₁H₂₇N₇O₁₄P₂
MDL Nummer: MFCD00036253
CAS nummer: 53-84-9

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR