Yeast extract, powder, ultrapure

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

J23547.A1
J23547.A1EA 73.6 EUR
J23547.A1
Yeast extract, powder, ultrapure
Media voor microbiologie Gedehydrateerde media voor microbiologie

  • Assay Protein (TN×6.25): 57 - 72%
  • Assay Salmonella: Negative
  • Assay Standard Plate Count: ≤15000/g
  • Residual Water(at Time of Packaging): ≤6%

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR