Masterflex® plugkranen met grote doorlaat, mannelijke Luer Lock, Avantor®
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MFLX30600-23EA 52.4 EUR
MFLX30600-23
Masterflex® plugkranen met grote doorlaat, mannelijke Luer Lock, Avantor®
Ventielen Ball Valves
Verbeter de vloeistofstroom met een grotere doorlaat dan standaard luer-stopkranen.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
More Product Information