β-Nicotinamide adenine dinucleotid, oxidised form (NAD, oxidised form)

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AE11.1 AE11.2 AE11.3
ROTHAE11.1EA 36.9 EUR
ROTHAE11.1 ROTHAE11.2 ROTHAE11.3
β-Nicotinamide adenine dinucleotid, oxidised form (NAD, oxidised form)
β-Nicotinamide adenine dinucleotid, oxidised form (NAD, oxidized form)
Formule: C₂₁H₂₇N₇O₁₄P₂
Smeltpunt: 140 to 142 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00036253
CAS nummer: 53-84-9
EINECS: 200-184-4
Merck Index: 13,06370

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR