Animal Component-Free Cell Dissociation Kit
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
05426
STMC05426EA 263 EUR
STMC05426
Animal Component-Free Cell Dissociation Kit
Celcultuursupplementen en -additieven
Dissociation kit for human stem and progenitor cells.
  • Optimised for the dissociation and passaging of human stem and progenitor cells
  • Obtain cleaner cultures with ready-to-use, animal component-free solutions
Use animal component-free dissociation kit for dissociating and passaging human stem and progenitor cells. Compatible with cells cultured in various media formulations, this kit comes with ready-to-use solutions, including ACF enzymatic dissociation solution and ACF enzyme inhibition solution.
Previously known as MesenCult™-ACF dissociation kit, this kit's name was changed to accommodate the use of this product with other cell types such as endothelial cells, HUVECs, mesenchymal cells, PSC-derived cells, mesenchymal stem and progenitor cells, and myogenic stem and progenitor cells.
Opgelet: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').