CellAdhere™ Type I Collagen, Human, Lyophilized
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
07005
STMC07005EA 664 EUR
STMC07005
CellAdhere™ Type I Collagen, Human, Lyophilized
Celcultuursupplementen en -additieven
Support cell attachment, growth, and proliferation with CellAdhere™ Type I Collagen, Human, Lyophilised. The most abundant type of collagen in the human body, type I collagen is a natural extracellular matrix protein that provides structural support and mimics physiological conditions, making it an ideal cultureware coating surface to support cell attachment in 2D applications.
CellAdhere™ Type I Collagen, Human, Lyophilised is prepared from an extracellular matrix secreted by normal human fibroblasts and is xeno-free. This sterility-tested product is purified using a highly controlled manufacturing process with validated steps to ensure the inactivation of possible viral contaminants. Endotoxin levels are 5,0 EU/ml or less, as assayed by Limulus amoebocyte lysate (LAL) assay. The optimal protein concentration may vary depending on the cell type being used, and therefore should be titrated for best results.
Order Now
Specificaties
Environmentally Preferable
Species Non-human primate,Human,Rat,Mouse
Bron