STEMdiff™ Neural Organoid Maintenance Kit
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
100-0120
STMC100-0120EA 315 EUR
STMC100-0120
STEMdiff™ Neural Organoid Maintenance Kit
Celcultuurmedia
Cell culture medium kit for long-term maintenance and maturation of patterned neural organoids.
Culture patterned brain organoids for extended periods (> 50 days) without the use of embedding matrices. With the essential components to prevent organoid fusion and retain a high yield of reproducible organoids, STEMdiff™ Neural Organoid Maintenance Kit is a serum-free cell culture medium designed for the maintenance and maturation of neural organoids adapted from protocols by Sergiu Paşca.
When used following STEMdiff™ Dorsal or Ventral Forebrain Organoid Differentiation Kits, STEMdiff™ Neural Organoid Maintenance Kit allows for long-term organoid experiments for neurodegeneration research, biomarker discovery, and toxicity assays. The resulting brain-region-specific organoids have a cellular composition and structural organization that is representative of the developing human forebrain. Organoids generated with these kits can also be co-cultured as assembloids to study brain region interactions.
Opgelet: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').