Anti-CD69 (AIM) Mouse Monoclonal Antibody [clone: FN50] (PE-Cy7)
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
100-1347
STMC100-1347EA 1034 EUR
STMC100-1347
Anti-CD69 (AIM) Mouse Monoclonal Antibody [clone: FN50] (PE-Cy7)
Antilichamen
Mouse monoclonal antibody against human CD69 (AIM).
  • B cells
  • Granulocytes
  • Subsets
  • Nk cells
  • T cells
The monoclonal mouse CD69 antibody reacts with CD69, a gp28/34 disulfide bonded homodimer with a molecular weight of 60 kDa under non-reducing conditions. CD69, also known as the Very Early Activation Antigen, EA-1, Leu23, Activation Inducer Molecule (AIM) and CLEC2C, is a type II transmembrane protein that is constitutively expressed by activated platelets. Use this monoclonal antibody in applications requiring detection of CD69 such as flow cytometry (More Product Information