RosetteSep™ HLA Myeloid Cell Enrichment Kit
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
15272HLA
STMC15272HLAEA 1230 EUR
STMC15272HLA
RosetteSep™ HLA Myeloid Cell Enrichment Kit
Media voor celscheiding
Immunodensity isolation of untouched myeloid cells for lineage-specific chimerism analysis.
  • Fast and easy-to-use
  • Requires no special equipment or training
  • Isolated cells are untouched
The RosetteSep™ HLA Myeloid Cell Enrichment Cocktail is designed to enrich myeloid (CD33+) cells from whole blood by negative selection for lineage-specific chimerism analysis. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognising CD3, CD8, CD19, CD56 and glycophorin A on red blood cells (RBCs). When centrifuged over a buoyant densityMore Product Information