Hausser Scientific™ Hemocytometer Cover Glasses
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
200-0641
STMC200-0641EA 182 EUR
STMC200-0641
Hausser Scientific™ Hemocytometer Cover Glasses
Dekglaasjes
Count cells with hemocytometer cover glasses designed specifically for use with the Hausser Scientific™ Bright-Line Hemocytometer.
  • Replacement cover glasses
These cover glasses facilitate manual cell counting at a fraction of the cost and footprint of automated cell counters.
Verpakking: They are available in packs of 10.
Opgelet: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or removeMore Product Information