Anti-CD4 Rat Monoclonal Antibody [clone: RM4-4] (FITC)
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
60029FI 60029FI.1
STMC60029FIEA 428 EUR
STMC60029FI STMC60029FI.1
Anti-CD4 Rat Monoclonal Antibody [clone: RM4-4] (FITC)
Antilichamen
Rat monoclonal IgG2b antibody against mouse CD4, FITC-conjugated.
  • T cells
  • T cells CD4
  • T cells regulatory
The RM4-4 antibody reacts with mouse CD4, an ~55 kDa single-chain type 1 transmembrane glycoprotein and member of the immunoglobin (Ig) superfamily; CD4 contains four extracellular Ig-like domains. CD4 is expressed at relatively high levels by most thymocytes and a subpopulation of T cells (T-helper/inducer cells), and at lower levels on dendritic cells. In the mouse, CD4 is not expressed by monocytes/macrophages. CD4 binds to a non-polymorphicMore Product Information