Anti-Igg2a Isotype Control Mouse Monoclonal Antibody [clone: MOPC-173] (Alexa Fluor® 488)

Leverancier: STEMCELL Technologies
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

60071AD
STMC60071ADEA 289 EUR
STMC60071AD
Anti-Igg2a Isotype Control Mouse Monoclonal Antibody [clone: MOPC-173] (Alexa Fluor® 488)
Antilichamen
Mouse monoclonal IgG2a, kappa isotype control antibody, Alexa Fluor® 488-conjugated.

The MOPC-173 antibody (IgG2a, kappa) is suitable for use as an isotype-matched control antibody in several applications to estimate the degree of non-specific binding by an antigen specific antibody. Ideally, the isotype control should have the same subclass of heavy chain (IgA, IgD, IgE, IgG, or IgM) and light chain (kappa or lambda) as the specific antibody being used. If a conjugated antibody is used, an isotype control conjugated to the same molecule (e.g. fluorochrome) should be chosen. The use of anMore Product Information
Order Now

Specificaties

Type antilichaam Igg2a Isotype Control
Environmentally Preferable
Clonality Monoklonaal
Conjugatie Alexa Fluor® 488
Clone MOPC-173
Reactivity
Host Mouse
Isotype IgG2a kappa

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR