RIPA Lysis and Extraction Buffer
Leverancier: G Bioscienes
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
786-489 786-490
786-489EA 240 EUR
786-489 786-490
RIPA Lysis and Extraction Buffer
Buffers Extractiebuffers
A complete lysis buffer for the release of cytoplasmic, membrane and nuclear proteins from adherent and suspension cultured mammalian cells. The RIPA lysis buffer is fully compatible with many applications, including reporter assays, protein assays, immunoassays and other protein purification techniques.