Benzonase® Nuclease
Leverancier: Merck Millipore (Novagen)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
70664-3 70746-3 70746-4 71206-3 71205-3
70664-3EA 313 EUR
70664-3 70746-3 70746-4 71206-3 71205-3
Benzonase® Nuclease
Enzymen
Benzonase® nuclease degrades all forms of DNA and RNA while having no proteolytic activity. Benzonase® is compatible with BugBuster® and PopCulture® protein extraction reagents and therefore can be added to these reagents to eliminate viscosity and remove nucleic acids from E. coli extracts.
  • Degrades all forms of DNA and RNA
  • Effective over a wide range of conditions
  • Exceptionally high specific activity
  • Ideal for removal of nucleic acids from recombinant proteins
  • Reduces viscosity, processing time and improves protein yields from cell extracts