VWR® 2XYT Medium Broth for Biotechnology
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
J902-100GEA 82.4 EUR
J902-100G J902-500G
VWR® 2XYT Medium Broth for Biotechnology
Media voor microbiologie Gedehydrateerde media voor microbiologie
2XYT medium broth powder is used to prepare rich broth for the propagation of M13 bacteriophage.
  • Save time by using pre-mixed, ready to use media
  • Homogenous, high quality powders
  • Light beige, free flowing powder
31 g of powder when dissolved per litre of deionised water will yield 16 g/L of casein peptone, 10 g/L of casamino acids and five g/L of sodium chloride.