Transfer membranes, PVDF, Owl™, Immobilon-P™
Leverancier: THERMO OWL SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TM151-1 TM151-4 TM151-5
OWLATM151-1EA 719 EUR
OWLATM151-1 OWLATM151-4 OWLATM151-5
Transfer membranes, PVDF, Owl™, Immobilon-P™
Transfermembranen
Hydrophobic polyvinylidene fluoride transfer membrane has a high signal-to-noise ratio, which allows for the use of mild blocking procedures
Membrane has high protein retention, is compatible with all standard visualization methods, and can be used for protein sequencing. Available in sheet or roll form.
Thermo Scientific offers Immobilon™-P, PVDF transfer membrane manufactured by Millipore™