E.Z.N.A.® RNALock stabilizer reagent
Leverancier: OMEGA BIO-TEK
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
R0424-01
R0424-02
R0424-01EA
81.7
EUR
R0424-01
R0424-02
E.Z.N.A.® RNALock stabilizer reagent
Buffers
Extractiebuffers
The E.Z.N.A.® RNALock Stabilizer Reagent is an innovative reagent which rapidly lyses cells and tissues to inactivate RNases at room temperature. Simply submerge the samples, such as cell cultures, animal tissues, or white blood cells into the RNALock Stabilizer Reagent. The intact cellular RNA will be preserved even at elevated temperature. Samples can be stored in this reagent in excess of 12 months at −20 °C.