Dehydrated media, Media fill
Leverancier: THERMO FISHER DIAGNOSTICSTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
A gamma-irradiated cold filterable vegetable peptone broth suitable for microbiological media fill trials (MFT) in the pharmaceutical industry. Cold Filterable Vegetable Peptone Broth (cfVPB) is a highly nutritious,
general purpose medium which supports the growth of a wide range of bacteria, yeasts and moulds.
- Easy to use
- Convient
- Animal free