Plating beads, ColiRollers™
Leverancier: Merck Millipore (Novagen)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
71013-3 71013-4
71013-3EA 44.4 EUR
71013-3 71013-4
Plating beads, ColiRollers™
Parels Glasparels
ColiRollers™ plating beads are a specially treated glass bead which offers a better way to perform plating bacteria for transformation and molecular cloning applications. ColiRollers™ beads eliminate the inconsistencies of conventional 'hockey stick' methods.
  • Non laborious and time saving
  • Entire stacks of plates can be processed at one time
  • Cells are evenly and completely spread over the agar surface
Leveringsinformatie: Each pack contains enough ColiRollers™ beads for 60 to 80 plates.