Pierce™ DNA Extraction Buffer, Phenol:Chloroform and Tris Buffer
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
17908 17909
PIER17908EA 122 EUR
PIER17908 PIER17909
Pierce™ DNA Extraction Buffer, Phenol:Chloroform and Tris Buffer
Buffers Extractiebuffers
Phenol-chloroform, is phenol supplied premixed with chloroform:isoamyl alcohol (25:24:1) and accompanying bottle of buffer to equilibrate at either pH 6,7 or pH 8,0.
  • Offers excellent, nuclease-free performance
  • Ideal for lower pH DNA extractions and for most DNA applications