Protein extraction reagents, CytoBuster™
Leverancier: Merck Millipore (Novagen)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
71009-3 71009-4
71009-3EA 126 EUR
71009-3 71009-4
Protein extraction reagents, CytoBuster™
Eiwitextractie Reagentia voor eiwitextractie
The CytoBuster™ protein extraction reagent is a proprietary formulation of detergents optimised for efficient extraction of soluble proteins from mammalian and insect cells. The gentle, non-ionic composition of the reagent enables isolation of functionally active endogenous or expressed proteins without a need for secondary treatment such as sonication or freeze/thaw.
CytoBuster™ protein extraction reagent has been specifically formulated for use in Western blotting, immunoprecipitation, and kinase/phosphatase assays. The reagent is compatible with protease inhibitors, kinase inhibitors, and phosphatase inhibitors.