Real-Time PCR kit for detection of Vibrio parahaemolyticus
Leverancier: Q BIOANALYTIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
QB-RTI-10-50 QB-RTI-10-1000 QBRTI-10-10SMP QB-RTI-10-250
710-1748EA 0 EUR
710-1748 710-1745 710-1746 710-1747
Real-Time PCR kit for detection of Vibrio parahaemolyticus
Testsystemen voor microbiologie PCR testen
Food safety experts like the German DGHM recommend to test warm water fish for Vibrio parahaemolyticus. With this Real-Time PCR test you have a very versatile method for food testing, which can gives you results within 24 hrs. The test was developed and validated within the German Network for Vibrio Research “VibrioNet” with many scientific institutions involved. It can be applied for food and sea water samples. In case of positive results you may be interested to determine whether or not the present strain has the ability to produce toxins (tdh/trh). You can use our Real-Time PCR Test for toxicity gene of Vibrio cholera for this purpose VWR catalogue number: QBIAQB-RTI-50. You can use this test in combination with an appropriate pre- enrichment in alkaline peptone-water or you can confirm bacterial colonies on agar plates.
  • TaqMan probes
  • Contains internal amplification control in HEX channel
  • Taget in FAM channel
  • Contains UNG to protect from re-amplification
  • Positive DNA and negative control included
Certificaten: DIN EN ISO 20838 conform detection in food, feed and water samples from pre-enrichment cultures or for confirmation of bacterial colonies